Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bub3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582108
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582108 Supplier Invitrogen™ Supplier No. PA582108
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YQTRCIRAFP NKQGYVLSSI EGRVAVEYLD PSPEVQKKKY AFKCHRLKEN NIEQIYPVNA ISFHNIHNTF ATGGSDGFVN IWDPFNKKRL CQFHRYPTSI ASLAFSNDGT TLAIASSYMY EMDDTEHPED GIFIRQVTDA ETKPKSP Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bub3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BUB3
Gene Accession No. O43684, Q9WVA3
Gene Alias Aa2-050; AU019800; AU021329; AU043350; AW146323; bub3; BUB3 budding uninhibited by benzimidazoles 3 homolog; BUB3 mitotic checkpoint protein; BUB3, mitotic checkpoint protein; BUB3L; Bub3-PA; Bub3-PB; budding uninhibited by benomyl; budding uninhibited by benzimidazoles 3 homolog; budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae); C78067; CG7581; CG7581-PA; CG7581-PB; dBUB3; Dmbub3; Dmel\CG7581; Dmel_CG7581; hBUB3; mitotic checkpoint component; Mitotic checkpoint protein BUB3; OTTHUMP00000020682; testicular tissue protein Li 27; WD repeat type I transmembrane protein A72.5
Gene Symbols BUB3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BUB3. Recombinant protein control fragment (Product #RP-102165).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12237, 9184
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.