Specifications
| Material | Borosilicate Glass |
| Type | Mineral Oil Bubbler |
| Includes | Ground Joint on the Bottom |
| Standard Taper Joint | 29/42, 24/40 |
| Length (Metric) | 157 mm |
| For Use With (Application) | Organic Synthesis |
| Quantity | 1 Each |