Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BubR1 Antibody (8G1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100353PE
Description
BubR1 Monoclonal specifically detects BubR1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
BubR1 | |
Monoclonal | |
PE | |
O60566 | |
BUB1B | |
A portion of human BubR1 tagged at the N-terminus with pMAL (residues 1-350). [UniProt# O60566] | |
Protein G purified | |
RUO | |
Primary | |
NB100-353 is specific for hBUBR1. There should be no cross-reactivity with hBUB1. | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
8G1 | |
Flow Cytometry | |
Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta | |
Mouse | |
120 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
701 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction