Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BubR1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100579R
Description
BubR1 Polyclonal antibody specifically detects BubR1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
BubR1 | |
Polyclonal | |
Western Blot | |
Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta | |
The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human budding uninhibited by benzimidazoles 1 homolog beta using the numbering given in TrEMBL entry Q8WV50 (GeneID 701). | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
701 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction