Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BubR1 Monoclonal Antibody (8G1), Active Motif™

Mouse Monoclonal Antibody
$135.30 - $405.90
Specifications
Antigen | BubR1 |
---|---|
Clone | 8G1 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2823
![]() ![]() |
Active Motif
39748 |
10 μg |
Each for $135.30
|
|
|||||
50-199-2822
![]() ![]() |
Active Motif
39747 |
100 μg |
Each for $405.90
|
|
|||||
Description
BUBR1 is a member of the Ser/Thr protein kinase family. It is a component of the mitotic checkpoint that delays anaphase until all chromosome are properly attached to the mitotic spindle. Compromised spindle checkpoints are thought to play an important role in genetic instability that predisposes cells to malignant transformations. Human cells contain two related checkpoint protein kinases, evolved from the BUB1 gene; hBUBR1 and hBUB1. They are important components of the spindle checkpoint Both kinases monitor mitotic kinetochore-microtubule interactions. hBUBR1 is essential for normal mitotic progression as it prevents cells from entering into anaphase too early. BUBR1 has been detected in human cancers. It has been identified as a novel binding partner of BCSG1 (Breast Cancer-Specific Gene 1), which has been suggested to accelerate the progression of breast cancer by compromising the mitotic checkpoint control through inactivation of BUBR1.Specifications
BubR1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
AU045529; BUB1 budding uninhibited by benzimidazoles 1 homolog beta; BUB1 mitotic checkpoint serine/threonine kinase B; Bub1A; BUB1B; BUB1B, mitotic checkpoint serine/threonine kinase; BUB1beta; BUBR1; budding uninhibited by benzimidazoles 1 beta; budding uninhibited by benzimidazoles 1 homolog beta; budding uninhibited by benzimidazoles 1 homolog, beta (S. cerevisiae); hBub1; hBUBR1; MAD3/BUB1-related protein kinase; MAD3L; Mitotic checkpoint kinase MAD3L; Mitotic checkpoint serine/threonine-protein kinase BUB1 beta; MVA1; Protein SSK1; SSK1 | |
BUB1B | |
IgG | |
Protein G | |
Antibody |
8G1 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O60566, Q9Z1S0 | |
12236, 701 | |
This BubR1 antibody was raised against recombinant protein corresponding to amino acids 1-350 of human BubR1. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
BUB1B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title