Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BubR1 Antibody (8G1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280606
Description
BubR1 Monoclonal specifically detects BubR1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.Specifications
BubR1 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:2000-1:5000, Flow Cytometry 1 ug per million cells, Immunocytochemistry/Immunofluorescence : 1:100, Immunoprecipitation : 1:10-1:500, CyTOF-ready | |
Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human and mouse. | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, CyTOF | |
8G1 | |
Unconjugated | |
PBS | |
BUB1B | |
A portion of human BubR1 tagged at the N-terminus with pMAL (residues 1-350). [UniProt# O60566] | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
701 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction