Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BUBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674792
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-792 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-792 Supplier Invitrogen™ Supplier No. PA112553
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

A suggested positive control for this product is mitotic NIH-3T3 cells.

BUBR1 is a member of the Ser/Thr protein kinase family. It is a component of the mitotic checkpoint that delays anaphase until all chromosome are properly attached to the mitotic spindle. Compromised spindle checkpoints are thought to play an important role in genetic instability that predisposes cells to malignant transformations. Human cells contain two related checkpoint protein kinases, evolved from the BUB1 gene; hBUBR1 and hBUB1. They are important components of the spindle checkpoint Both kinases monitor mitotic kinetochore-microtubule interactions. hBUBR1 is essential for normal mitotic progression as it prevents cells from entering into anaphase too early. BUBR1 has been detected in human cancers. It has been identified as a novel binding partner of BCSG1 (Breast Cancer-Specific Gene 1), which has been suggested to accelerate the progression of breast cancer by compromising the mitotic checkpoint control through inactivation of BUBR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BUBR1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene BUB1B
Gene Accession No. Q9Z1S0
Gene Alias AU045529; BUB1 budding uninhibited by benzimidazoles 1 homolog beta; BUB1 mitotic checkpoint serine/threonine kinase B; Bub1A; Bub1b; BUB1B, mitotic checkpoint serine/threonine kinase; BUB1beta; BUBR1; budding uninhibited by benzimidazoles 1 beta; budding uninhibited by benzimidazoles 1 homolog beta; budding uninhibited by benzimidazoles 1 homolog, beta (S. cerevisiae); hBub1; hBUBR1; MAD3/BUB1-related protein kinase; MAD3L; Mitotic checkpoint kinase MAD3L; mitotic checkpoint serine/threonine-protein kinase BUB1 beta; MVA1; Protein SSK1; SSK1
Gene Symbols BUB1B
Host Species Sheep
Immunogen Recombinant protein corresponding to residues 232-603 of mouse Bub1b.
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12236
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.