Learn More
BubR1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300386AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Compromised spindle checkpoints are thought to play an important role in genetic instability that predisposes cells to maligt transformations. Human cells contain two related checkpoint protein kinases, evolved from the BUB1 gene; hBUBR1 and hBUB1. They are important components of the spindle checkpoint Both kinases monitor mitotic kinetochore-microtubule interactions. hBUBR1 is essential for normal mitotic progression as it prevents cells from entering into anaphase too early. BUBR1 has been detected in human cancers. It has been identified as a novel binding partner of BCSG1 (Breast Cancer-Specific Gene 1), which has been suggested to accelerate the progression of breast cancer by compromising the mitotic checkpoint control through inactivation of BUBR1.Specifications
| BubR1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O60566, Q9Z1S0 | |
| Bub1b | |
| Between 350 and 400 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| BUB1B | |
| BUB1B, Budding Uninhibited by Benzimidazoles 1 beta, hBUBR1, MAD3L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12236, 171576, 701 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.