Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Boiling Point Tubes and Capillaries
SDP

Catalog No. 05402814 Shop All BUCHI Products
Click to view available options
Product Type:
Boiling Point Capillaries
Boiling Point Tubes
Packaging Quantity:
10/Pk.
100/Pk.

Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.

  • High precision for reliable results
  • Meets Siwoloboff and Eu Pharmacopeia standards
  • Designed for BUCHI melting and boiling point instruments
  • Compatible with other instruments

Specifications

Product Type Boiling Point Tubes
Diameter (Metric) 2.75 mm
For Use With (Application) For reproducible boiling point determinations
For Use With (Equipment) Boiling point apparatus
Material Borosilicate Glass 3.3
Packaging Quantity 10/Pk.
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.