Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments
SDP

Catalog No. 10987120 Shop All BUCHI Products
Click to view available options
Product Type:
Calibration Kit
Calibration Set
Verification Kit
For Use With (Equipment):
BUCHI Melting Point Instrument
Melting Point Instruments

Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.

Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.

  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C

Specifications

Product Type Calibration Set
For Use With (Equipment) BUCHI Melting Point Instrument
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.