Learn More
Thermo Scientific™ Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
GSA_VA
Catalog No. p-8872000
Click to view available options
Type:
Sealed Bucket
Unsealed Bucket
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Max Tube Dimensions: Height <33 mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Capacity 4 Standard or 2 Midi-Deepwell plates
Type Unsealed Bucket
Description Unsealed Buckets (set of 2)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.