Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BUD13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558351
Description
Immunogen sequence: PGHQDSDSDL SPPRNRPRHR SSDSDLSPPR RRQRTKSSDS DLSPPRRSQP PGKKAAHMYS GAKTGLVLTD IQREQQELKE QDQETM Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 81%.
BUD13 is a protein coding gene involved in pre-mRNA splicing as component of the activated spliceosome.
Specifications
BUD13 | |
Polyclonal | |
Unconjugated | |
BUD13 | |
AV305342; BUD13; BUD13 homolog; BUD13 homolog (yeast); Cwc26; D030060M11Rik; fSAP71; functional spliceosome-associated protein 71 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
215051, 84811 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8R149, Q9BRD0 | |
BUD13 | |
Recombinant protein corresponding to Human BUD13. Recombinant protein control fragment (Product #RP-97347). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction