Learn More
BUD13 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303321AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
BUD13 is a protein coding gene involved in pre-mRNA splicing as component of the activated spliceosome.Specifications
BUD13 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9BRD0 | |
BUD13 | |
Between 150 and 200 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
BUD13 | |
fSAP71, BUD13 homolog, functional spliceosome-associated protein 71 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84811 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.