Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BUD31 Monoclonal Antibody (1E10)
GREENER_CHOICE

Catalog No. PIMA520551
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520551 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520551 Supplier Invitrogen™ Supplier No. MA520551
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: EPHEGKRKVE SLWPIFRIHH QKTRYIFDLF YKRKAISREL YEYCIKEGYA DKNLIAKWKK QGYENLCCLR CIQTRDTNFG TNCICRVPKS KLEVGRIIEC THCGCRGCSG.

G10 maternal transcript homolog, G10 protein, functional spliceosome-associated protein 17, maternal G10 transcript.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BUD31
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E10
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Bud31
Gene Accession No. P41223
Gene Alias Bud31; BUD31 homolog; BUD31 homolog (S. cerevisiae); BUD31 homolog (yeast); C77604; Cwc14; EDG2; EDG-2; fSAP17; functional spliceosome-associated protein 17; G10; G10 maternal transcript homolog; maternal G10 transcript; protein BUD31 homolog; Protein EDG-2; protein G10 homolog; YCR063W
Gene Symbols Bud31
Host Species Mouse
Immunogen BUD31 (NP_003901, 35 a.a. ~ 144 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8896
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.