Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BUD31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549804 Supplier Invitrogen™ Supplier No. PA549804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total BUD31 protein.

G10 maternal transcript homolog, G10 protein, functional spliceosome-associated protein 17, maternal G10 transcript.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BUD31
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Bud31
Gene Accession No. O70454, P41223, Q6PGH1
Gene Alias Bud31; BUD31 homolog; BUD31 homolog (S. cerevisiae); BUD31 homolog (yeast); C77604; Cwc14; EDG2; EDG-2; fSAP17; functional spliceosome-associated protein 17; G10; G10 maternal transcript homolog; maternal G10 transcript; protein BUD31 homolog; Protein EDG-2; protein G10 homolog; YCR063W
Gene Symbols Bud31
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human BUD31.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231889, 8896, 89819
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.