Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BUD31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598036
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598036 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598036 Supplier Invitrogen™ Supplier No. PA598036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BUD31 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

G10 maternal transcript homolog, G10 protein, functional spliceosome-associated protein 17, maternal G10 transcript.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BUD31
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Bud31
Gene Accession No. P41223, Q6PGH1
Gene Alias Bud31; BUD31 homolog; BUD31 homolog (S. cerevisiae); BUD31 homolog (yeast); C77604; Cwc14; EDG2; EDG-2; fSAP17; functional spliceosome-associated protein 17; G10; G10 maternal transcript homolog; maternal G10 transcript; protein BUD31 homolog; Protein EDG-2; protein G10 homolog; YCR063W
Gene Symbols Bud31
Host Species Rabbit
Immunogen Recombinant Human Protein BUD31 homolog protein (1-144AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231889, 8896
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.