Learn More
Invitrogen™ BUD31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120148
Description
Positive test controls include: Raji, Jurkat, HeLa, 293T, Mouse brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: MPKVKRSRKA PPDGWELIEP TLDELDQKMR EAETEPHEGK RKVESLWPIF RIHHQKTRYI FDLFYKRKAI SRELYEYCIK EGYADKNLIA KWKKQGYENL CCLRCIQTRD TNFGTNCICR VPKSKLEVGR IIECTHCGCR GCSG.
G10 maternal transcript homolog, G10 protein, functional spliceosome-associated protein 17, maternal G10 transcript.
Specifications
BUD31 | |
Polyclonal | |
Unconjugated | |
Bud31 | |
Bud31; BUD31 homolog; BUD31 homolog (S. cerevisiae); BUD31 homolog (yeast); C77604; Cwc14; EDG2; EDG-2; fSAP17; functional spliceosome-associated protein 17; G10; G10 maternal transcript homolog; maternal G10 transcript; protein BUD31 homolog; Protein EDG-2; protein G10 homolog; YCR063W | |
Rabbit | |
Affinity Chromatography | |
RUO | |
231889, 8896, 89819 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.09 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O70454, P41223, Q6PGH1 | |
Bud31 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human BUD31 (NP_003901.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.