Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUD31 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 117981AP150UL

Description
G10 maternal transcript homolog, G10 protein, functional spliceosome-associated protein 17, maternal G10 transcript.Specifications
BUD31 | |
Polyclonal | |
Unconjugated | |
BUD31 | |
BUD31, BUD31 homolog (S. cerevisiae), EDG 2, EDG2, fSAP17, G10, Protein BUD31 homolog, Protein EDG 2, Protein G10 homolog, YCR063W | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231889, 8896, 89819 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70454, P41223, Q6PGH1 | |
BUD31 | |
BUD31 Fusion Protein Ag2373 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction