Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chemical Solution, Buffer Solution pH 10.0 at 25°C, Color Coded Blue, Traceable to NIST, LabChem
Click to view available options
Quantity:
500 mL
4 L
20 L
Description
- Chemical Solution, Buffer Solution.
- pH 10.0 at 25°C.
- Color Coded Blue.
- Traceable to NIST.
Specifications
Specifications
CAS | 26628-22-8, 1310-73-2, 497-19-8, 7732-18-5, 144-55-8 |
Physical Form | Aqueous Solution |
Quantity | 4 L |
Solution Type | Buffer |
Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction