Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 7, Color-coded Yellow (Certified), Fisher Chemical™ PROMO

Catalog No. SB107500S24 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Color-coded buffer solution for research use.

  • Color-coded for your convenience
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution, pH 7, Color-coded Yellow
Melting Point 0°C
CAS 7778-77-0, 1310-73-2, 7732-18-5, 1934-21-0
Boiling Point 100°C
Color Yellow
Density 1 g/cm3
pH 7
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00134130
Packaging Poly Bottle
Specific Gravity 1
Traceability to NIST Yes
Vapor Pressure 760 mmHg at 20°C
Solubility Information Soluble in Water
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.