Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 10, Color-coded Blue (Certified), Fisher Chemical™ PROMO

Catalog No. SB115500S24 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL

Color-coded buffer solution for research use.

  • Color-coded for your convenience
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution, pH 10, Color-coded Blue
Melting Point 0°C
Boiling Point 100°C
Color Blue
CAS 6381-92-6, 584-08-7, 1332-77-0, 1310-58-3
pH 10
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
Packaging Poly Bottle
MDL Number MFCD00146206
Specific Gravity 1.013 at 25°C
Traceability to NIST Yes
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.