Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution, pH 4, Color-coded Red (Certified), Fisher Chemical™ PROMO
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Description
- pH/temperature conversions given on label

Specifications
Specifications
Chemical Name or Material | Buffer Solution, pH 4, Color-coded Red |
Melting Point | 0°C |
CAS | 50-00-0, 67-56-1, 877-24-7, 7732-18-5, 16423-68-0 |
Boiling Point | 100°C |
Color | Red |
Density | 1 g/cm3 |
pH | 4 |
Physical Form | Liquid |
Quantity | 500 mL |
Identification | Pass Test |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction