Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Concentrate, pH 4.00 (Certified), Fisher Chemical™

Catalog No. p~213027 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB99-500 500 mL
SB99-1 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB99-500
Only null left
Add to Cart

Specifications

Chemical Name or Material Buffer Concentrate, pH 4.00
Melting Point 0°C
CAS 877-24-7, 50-00-0, 67-56-1, 7732-18-5
Boiling Point 100°C
Color Undesignated
Density 1.0g/cm³
pH 4.0
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00013070
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Vapor Pressure 14mmHg at 20°C
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.