Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer concentrate pH 4.00, For 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution, Honeywell Fluka™

Catalog No. p-7122126
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.

for 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer concentrate
pH 4
Quantity 1 Pc.
Packaging Ampoule
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.