Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer HF-Improved, Thermo Scientific Chemicals

Catalog No. p-4407181
Click to view available options
Quantity:
250 mL
1 L

Used For Pretreatment of Planar Silicon Devices When Plating With Nickel Plating Solution Products 44069 and 44070

Used for pretreatment of planar silicon devices when plating with Nickel plating solution products 44069 and 44070.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer HF-Improved
Boiling Point 103°C to 105°C
Color Colorless
Density 1.12
Physical Form Liquid
Quantity 250 mL
MDL Number MFCD00132734

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.