Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 2.00 ± 0.01 at 25°C Colored Red, Reagents

Catalog No. 01337440
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-440 20 L
01-337-439 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-440 Supplier Reagents Holdings Llc Supplier No. C215540R20F
Only null left
Add to Cart
Add to Cart

Buffer pH 2.00 ± 0.01 at 25°C Colored Red, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Buffer pH 2.00 ± 0.01 at 25°C Colored Red
CAS 7732-18-5, 7447-40-7, 7647-01-0
Melting Point 0°C
Boiling Point 100°C
Color Red
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 2
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 1.001
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.