Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 4.00± 0.01 at 25°C Reference Standard, Reagents

Catalog No. 01337426
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L

Catalog No. 01-337-426

Supplier: Reagents Holdings Llc C2155510500A

Only null left
Add to Cart
Add to Cart

Buffer pH 4.00± 0.01 at 25°C Reference Standard, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.