Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 5.50 ± 0.01 at 25°C Reference Standard, Reagents

Catalog No. 01337425
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-425 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-425 Supplier Reagents Holdings Llc Supplier No. C21555401A
Only null left
Add to Cart
Add to Cart

Buffer pH 5.50 ± 0.01 at 25°C Reference Standard, NIST Traceable, use for pH meter calibration. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.