Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Precision Reference Standard, pH 10.000 ± 0.005 at 25°C, Ricca Chemical
SDP

Catalog No. 160216
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Standardized against NIST-certified pH references. High-Accuracy pH Buffer solutions are for use with meters with 0.001 resolution.

  • High-accuracy liquid 10.000 pH buffer
  • Sold in individual bottles of 500 mL, or as a case of twelve.
  • Store away from direct sunlight

Specifications

Chemical Name or Material Buffer, Precision Reference Standard
Name Note pH 10.000 ± 0.005 at 25 deg C (Color Coded Blue)
Melting Point 0°C
CAS 497-19-8, 7732-18-5, 1310-73-2
Boiling Point 100°C
Color Blue
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 10
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Clear PET-G Bottle
Solubility Information Miscible
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.