Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C, Ricca Chemical

Shop All Ricca Chemical Company Products


Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | Buffer, Reference Standard |
Name Note | pH 10.00 ± 0.01 at 25 deg C |
CAS | 497-19-8, 144-55-8, 1310-73-2, 7732-18-5 |
CAS Min % | 97.0, 0.51, 0.39, 0.03 |
CAS Max % | 100.0, 0.53, 0.41, 0.03 |
Concentration | 1X |
For Use With (Application) | Calibration |
For Use With (Equipment) | pH Meter and Electrode |
pH | 10 |
Physical Form | Liquid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction