Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 11.00 ± 0.01 at 25°C, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Clear PET-G Bottle
Cubitainer
Natural Poly Bottle
Specifications
Specifications
| Chemical Name or Material | Buffer, Reference Standard |
| Name Note | pH 11.00± 0.01 at 25°C |
| Melting Point | 0°C |
| CAS | 7558-79-4, 1310-73-2, 7732-18-5 |
| Boiling Point | 100°C |
| CAS Min % | 97.0, 1.02, 0.11 |
| Color | Colorless |
| CAS Max % | 100.0, 1.06, 0.11 |
| Concentration | 1X |
| For Use With (Application) | Calibration |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction