Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 6.00 ± 0.01 at 25°C, Ricca Chemical
SDP

Catalog No. p-3251432 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack

Specifications

Chemical Name or Material Buffer, Reference Standard, pH 6.00 ± 0.01 at 25°C
Melting Point ∼0°C
CAS 7778-77-0, 7558-79-4, 7732-18-5, 1310-73-2, PP26628-22-8
Boiling Point ∼100°C
CAS Min % 99.08, 0.81, 0.1, 0.01, 0.0028
Color Colorless
CAS Max % 99.08, 0.81, 0.1, 0.01, 0.0028
Density 1
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 6
Physical Form Liquid
Quantity 20 x 20 mL
Solution Type pH Buffer
Packaging Pack
Solubility Information Miscible in water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.