Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 7.38 ± 0.01 at 25°C, Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
4 L
Specifications
Specifications
Chemical Name or Material | Buffer, Reference Standard |
Name Note | pH 7.38± 0.01 at 25°C |
CAS | 7558-79-4, 7778-77-0, 7732-18-5 |
CAS Min % | 97.0, 0.86, 0.29 |
Color | Colorless |
CAS Max % | 100.0, 0.9, 0.3 |
Concentration | 1X |
For Use With (Application) | Calibration |
For Use With (Equipment) | pH Meter and Electrode |
pH | 7.38 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction