Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 8.00 ± 0.01 at 25°C, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
Specifications
Specifications
| Chemical Name or Material | Buffer, Reference Standard |
| Name Note | pH 8.00± 0.01 at 25°C |
| CAS | 7778-77-0, 7558-79-4, 7732-18-5 |
| Color | Colorless |
| Concentration | 1X |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 8 |
| Physical Form | Liquid |
| Quantity | 20 L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction