Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 9.00 ± 0.01 at 25°C, Ricca Chemical
SDP

Catalog No. p-3251440 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R159000020 20 x 20 mL Pack
1590-16 500 mL Natural Poly Bottle
1590-32 1 L Natural Poly Bottle
1590-1 4 L Natural Poly Bottle
1590-25 10 L Cubitainer
R15905 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R159000020 Supplier Ricca Chemical Company Supplier No. R159000020PK
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Buffer, Reference Standard, pH 9.00 ± 0.01 at 25°C
Melting Point 0°C
CAS 10043-35-3, 7447-40-7, 10043-35-3, 1310-73-2, PP26628-22-8
Boiling Point 100°C
CAS Min % 99.22, 0.37, 0.31, 0.09, 0.01
Color Colorless
CAS Max % 99.22, 0.37, 0.31, 0.09, 0.01
Density 1
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 9
Physical Form Liquid
Quantity 20 x 20 mL
Solution Type pH Buffer
Packaging Pack
Solubility Information Miscible in water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.