Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 9.00 ± 0.01 at 25°C, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
Specifications
Specifications
Chemical Name or Material | Buffer, Reference Standard |
Name Note | pH 9.00± 0.01 at 25°C |
CAS | 10043-35-3, 7447-40-7, 1310-73-2, 7732-18-5 |
Color | Colorless |
Concentration | 1X |
For Use With (Application) | Calibration |
For Use With (Equipment) | pH Meter and Electrode |
pH | 9 |
Physical Form | Liquid |
Quantity | 4 L |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction