Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 4.0 (20°C), Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide, Solstice

Catalog No. 6002469 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
6 x 500 mL
10 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-024-69 5 L
60-024-68 500 mL
60-024-67 1 L
60-046-294 6 x 500 mL
60-046-293 6 x 1 L
60-024-66 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-024-69 Supplier Solstice Supplier No. 336655LVP
Add to Cart
Add to Cart

Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

pH 4.0
Quantity 5 L
UN Number NONH for all modes of transport
IUPAC Name amine hydrate
Chemical Name or Material Buffer solution
Hazard Statement P280-P305 + P351 + P338-P337 + P313
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.