Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution A, for Sulfate Analysis (Turbidimetric Method), Ricca Chemical
SDP

Catalog No. p-3251407
Click to view available options
Quantity:
250 mL
1 L

For Analysis of Sulfate (Turbidimetric Method)

Specifications

Color Colorless
Packaging Type Natural Poly Bottle
Grade Chemical Solution
Melting Point ∼ 0°C
Product Type Buffer Solution
Quantity 1 L
Solution Type Buffer
For Use With (Application) Sulfate Analysis (Turbidimetric Method)
Reagent Type Buffer Solution
Form Liquid
Format Poly Bottle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.