Learn More
Buffer Solution, pH 1.68 +/-0.02 at 25°C, Reference Standard, Spectrum™ Chemical

Description
Spectrum™ Chemical Buffer Solution, pH 1.68 ± 0.02 at 25°C, Reference Standard is an inorganic solution in water. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum's own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.
Specifications
Specifications
CAS | 7647-01-0, 7447-40-7, 7732-18-5 |
CAS Max % | 1%, 98% |
For Use With (Application) | Titration |
For Use With (Equipment) | pH Meter |
pH | 1.66 to 1.70 |
Physical Form | Aqueous Solution |
Quantity | 4 L |
Solution Type | Standard |
Packaging | Poly Bottle |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.