Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 1.68 +/-0.02 at 25°C, Reference Standard, Spectrum™ Chemical
SDP

Catalog No. p-7157062 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
Packaging:
Poly Bottle

B-220

Spectrum™ Chemical Buffer Solution, pH 1.68 ± 0.02 at 25°C, Reference Standard is an inorganic solution in water. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum's own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Specifications

CAS 7647-01-0, 7447-40-7, 7732-18-5
CAS Max % 1%, 98%
For Use With (Application) Titration
For Use With (Equipment) pH Meter
pH 1.66 to 1.70
Physical Form Aqueous Solution
Quantity 500 mL
Solution Type Standard
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.