Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 11.0 at 25°C, Certified, LabChem™

Catalog No. p-5379472
Click to view available options
Quantity:
500 mL
1 L
4 L

Specifications

Chemical Name or Material Buffer Solution pH 11.0 at 25°C
Color Colorless
CAS Max % 99.64, 0.35, 0.01
CAS 7558-79-4, 1310-73-2
Decomposition Information Phosphine; Phosphorus oxides; Phosgene
Density 1g/mL
pH 11.00 ±0.05
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula Na2HPO4
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 141.96
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.