Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution, pH 13.0, Reference Standard, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
13.00 ±0.05
4 L
Description
Spectrum™ Chemical Buffer Solution, pH 13.0, Reference Standard
Specifications
Specifications
| CAS | 1310-73-2 |
| CAS Max % | 100% |
| pH | 13.00 ±0.05 |
| Quantity | 13.00 ±0.05 |
| Identification | pH Measurement |
| Packaging | Poly Bottle |
| InChI Formula | pH Meter |
| PubChem CID | Poly Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction