Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 2.00 +/-0.02 at 25°C, Reference Standard, Spectrum™ Chemical
SDP

Catalog No. 18601126
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle

B-225

Spectrum™ Chemical Buffer Solution, pH 2.00 ± 0.02 at 25°C, Reference Standard is a mixture of weak acid and its conjugate base in order to prevent changes in the pH of a solution. Buffer solutions let you calibrate any pH tester to ensure accurate readings when measuring pH in water samples and process liquids.

Specifications

CAS 7647-01-0, 7447-40-7, 7732-18-5
CAS Max % 1%, 98%
For Use With (Application) pH Measurement
For Use With (Equipment) pH Meter
pH 2.00 ±0.02
Physical Form Aqueous Solution
Quantity 4 L
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.