Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution, pH 2.00 +/-0.02 at 25°C, Reference Standard, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle
Description
Spectrum™ Chemical Buffer Solution, pH 2.00 ± 0.02 at 25°C, Reference Standard is a mixture of weak acid and its conjugate base in order to prevent changes in the pH of a solution. Buffer solutions let you calibrate any pH tester to ensure accurate readings when measuring pH in water samples and process liquids.
Specifications
Specifications
CAS | 7647-01-0, 7447-40-7, 7732-18-5 |
CAS Max % | 1%, 98% |
For Use With (Application) | pH Measurement |
For Use With (Equipment) | pH Meter |
pH | 2.00 ±0.02 |
Physical Form | Aqueous Solution |
Quantity | 500 mL |
Packaging | Poly Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction