Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 2.00 (Certified), Fisher Chemical™
GSA_VA

Catalog No. p-212999 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Poly Bottle
PolyPac™

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution, pH 2.00
Melting Point 0°C
CAS 7447-40-7, 7647-01-0, 50-00-0, 67-56-1, 7732-18-5
Boiling Point 100°C
Color Undesignated
Density 1.0g/cm³
pH 2.00
Physical Form Liquid
Quantity 500 mL
Identification In Pass Test
Packaging Poly Bottle
MDL Number MFCD00146206
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.