Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 3.0 (20°C), Citric acid / sodium hydroxide / sodium chloride solution, with fungicide, traceable to SRM from NIST and PTB, Honeywell Fluka™

Catalog No. p-7122050
Click to view available options
Quantity:
1 L
6 x 1 L

Citric acid / sodium hydroxide / sodium chloride solution, with fungicide, traceable to SRM from NIST and PTB

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 3.0 (20°C)
Name Note Citric acid / sodium hydroxide / sodium chloride solution, with fungicide, traceable to SRM from NIST and PTB
CAS 1310-73-2, 77-92-9, 7647-14-5, 7732-18-5
CAS Min % ≥98.0000%
CAS Max % ≥98.0000%
Density 1.000 g/cm3
Quantity 6 x 1 L
Packaging Poly Bottle
UN Number NONH for all modes of transport
Grade puriss. p.a.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.