Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 3.00 (Certified), Fisher Chemical™
GSA_VA

Catalog No. p~213006 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB97-500 500 mL Poly Bottle
SB97-20 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SB97-500
Only null left
Add to Cart
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution, pH 3.00
Melting Point 0°C
CAS 877-24-7, 7647-01-0, 50-00-0, 67-56-1, 7732-18-5
Boiling Point 100°C
Color Undesignated
Density 0.7g/cm³
pH 3.00
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
Packaging Poly Bottle
MDL Number MFCD00146206
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.