Learn More
Buffer Solution pH 7.0 (20°C), Potassium Dihydrogen Phosphate / Disodium Hydrogen Phosphate, Traceable to SRM from NIST, with Fungicide, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
Leading the Industry For Over 65 Years. Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured.
Composition per liter:
- 3.52 g Potassium dihydrogen phosphate (KH2PO4)
- 7.26 g di-Sodium hydrogen phosphate-2-hydrate (Na2HPO4*2H20)
Specifications
Specifications
| pH | 7.0 |
| Quantity | 5 L |
| MDL Number | MFCD00066650 |
| UN Number | NONH for all modes of transport |
| IUPAC Name | amine hydrate |
| Chemical Name or Material | Buffer solution |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.