Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 7.0 at 25°C, Color Coded Yellow, Certified, LabChem™
GSA_VA

Catalog No. p-5379483
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer™
Pack
Poly Bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material Buffer Solution pH 7.0 at 25°C
Name Note Color Coded Yellow
Color Yellow
CAS Max % 99.24, 0.68, 0.08
CAS 7778-77-0, 1310-73-2
Decomposition Information Phosphorus oxides; Sodium oxide
pH 7.00 ±0.01
Physical Form Liquid
Quantity 20 x 20 mL
Identification Passes Test
Linear Formula KH2PO4
Packaging Pack
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.09
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.