Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution pH 7.38 at 25°C, Certified, LabChem™
Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Buffer Solution pH 7.38 at 25°C |
Color | Colorless |
CAS Max % | 99.22, 0.68, 0.1 |
CAS | 7778-77-0, 1310-73-2 |
Decomposition Information | Phosphorus oxides; Sodium oxide |
pH | 7.38 ±0.01 |
Physical Form | Liquid |
Quantity | 500 mL |
Identification | Passes Test |
Linear Formula | KH2PO4 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction